Cole-Parmer TC FLASK-ST-SW/PLUG 50ML 200/CS
Representative image only.
Cole-Parmer TC FLASK-ST-SW/PLUG 50ML 200/CS
Cole-Parmer | Item: 1597124
Optimize the cultivation of cells and tissues in your lab with Cole-Parmer flasks
₹16,600.00INR / Case of 1
₹16,600.00INR / Case of 1
Product Description
Key Features
  • Designed for consistency, cell health and reproducibility, with surface and ancillary options
  • Innovative angled neck design offers easy pipetting and cell scraper access
  • Select from non-treated and surface-treated flasks with growth areas ranging from 12.5 to 300cm2
  • Corrugation free and smooth flask surface maximizes usable growth area
  • Upper triangular and wider base shape provides stability
  • Protuding ridges on the back side of the flask makes it easy for stacking
  • Gamma sterilized with strict integrity testing
  • Certified DNase-, RNase-free, and non-pyrogenic
  • Graduation marked on both sides along with special area near neck for marking
  • 2 different cap styles can be used in both open and closed system
  • Plug sealing caps - Standard polyethylene caps providing liquid and gas shear seal
  • Vented caps - Vented polyethylene caps contain a 0.22μ hydrophobic filter to allow gas exchange and minimize risk of cross contamination
  • Packaged in durable zip re-sealable self-standing plastic bags that allow flasks to remain upright and reduce contamination
  • Inner packaging is individually labelled for lot traceability
Sterile
Specifications
  • Capacity (mL)50
  • Cap TypeStandard
  • Culture Area (cm²)25
  • DescriptionStandard, surface treated