Cole-Parmer® Carboy w/ Handles, LDPE
Integrated handles make transporting solutions easy and safe
  • Leakproof PP cap
  • Easy to read, molded graduations
These rugged, durable plastic carboys allow you to meet your large volume liquid storage and dispensing needs. Integrated wide handles make them easy to carry and pour, even with gloves on. Polypropylene (PP) cap with integrated seal ring ensures leakproof seal. The carboys maintain flexibility and shatterproof safety down to -103°F (-75°C).
13,200.00 - ₹37,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
10 83 mm Low Density Polyethyene (LDPE)
₹13,400.00INR / Each
COMPARE
20 83 mm Low Density Polyethyene (LDPE)
₹20,400.00INR / Each
COMPARE
50 83 mm Low Density Polyethyene (LDPE)
₹37,000.00INR / Each
COMPARE
10 100 mm Low Density Polyethyene (LDPE)
₹13,200.00INR / Each
COMPARE
20 100 mm Low Density Polyethyene (LDPE)
₹20,100.00INR / Each