Cole-Parmer® Burette (Buret) and Stand Kit
Bundled for convenience and to save you money!
  • Class B burette (buret) meets your basic measuring and dispensing requirements — complete with burette clamp and lab stand
Handy kit includes a nickel-plated zinc, double-burette (buret) clamp; clamp holder; and lab stand with aluminum rod and chemical-resistant porcelain base. The burette clamp can hold up to two burettes—one 50-mL burette is included.
Our Class B 50-mL burette is manufactured from high-quality borosilicate glass, and designed for accuracy and durability. Burette features a carefully drawn tip for a reliable drainage rate, a noncontaminating, 2-mm, straight-bore PTFE stopcock, and brown-colored permanent graduations.
54,500.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Burette and Burette Stand Kit; Burette, Clamp, and Stand
₹54,500.00INR / Each