Cole-Parmer Built-In Glass Prefilters with Final Filter
Borosilicate glass fiber prefilters combined with cellulose acetate membranes extend the life of the final filter when using viscous, proteinaceous, or particulate-laden samples
  • Use these filters to eliminate prefiltration step, reduce retention volume and sample loss, and achieve faster flow rates
Filters are Triton-free and have a bidirectional membrane support and luer lock inlet/outlet for a secure fit to the syringe. Membrane type and pore size are printed on each filter for easy identification. All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. Sterile filters are sterilized by gamma radiation. They are certified nonpyrogenic, noncytotoxic, and are blister packed.
18,200.00 - ₹35,900.00INR / Box of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
COMPARE
28 0.2 Yes
₹18,200.00INR / Box of 50
COMPARE
28 0.2 No
₹34,900.00INR / Box of 100
COMPARE
28 0.45 No
₹35,900.00INR / Box of 100
COMPARE
28 0.8 No
₹34,900.00INR / Box of 100