Cole-Parmer® Broad-Spout Heatable PTFE Beakers
Stand up to the harshest chemicals
  • Pour easily with broad spout
  • Autoclavable
These high-purity PTFE beakers are ideal for use with the harshest chemicals, including caustics. Heatable beakers are made for use on a hot plate—the single-molded, specially formulated PTFE-carbon base offers excellent heat transfer and can be heated to 518°F (270°C) without distortion.
16,500.00 - ₹41,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Height (in)
Availability Pricing
COMPARE
100 Polytetrafluoroethylene (PTFE) 2 7/8
₹16,500.00INR / Each
COMPARE
250 Polytetrafluoroethylene (PTFE) 3 3/4
₹20,500.00INR / Each
COMPARE
400 Polytetrafluoroethylene (PTFE) 4 1/2
₹41,800.00INR / Each