Cole-Parmer® Bottletop Dispenser with Calibration
Clean, safe and precise dispensing at very affordable prices
  • Set and lock volume for optimized accurate results
  • Available in six volume ranges from 0.25 mL to 100 mL
  • Fully autoclavable and superior chemical compatibility
  • Jam resistant with smooth plunger movement
Cole-Parmer’s bottletop dispenser offers great performance over long period of time. Spring-less technology enables customer to use both organic and inorganic reagents. Jam-free piston not only helps for free piston movement but also ensures service-free operation. For flexibility of work the adapter is designed to rotate 360° which are available in various sizes - 28mm, 32mm, 36mm, 40mm and 45mm. The PTFE piston with ETP O-ring provide frictionless movement reducing the occurrence of bubbles in your dispenses.
25,100.00 - ₹45,300.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Dispense Volume (mL) Min Dispense Volume (mL) Description
Availability Pricing
COMPARE
2.5 0.25 Bottletop Dispenser, without Recirculation Valve; 0.25 to 2.5 mL
₹25,100.00INR / Each
COMPARE
5 0.5 Bottletop Dispenser, without Recirculation Valve; 0.5 to 5 mL
₹25,100.00INR / Each
COMPARE
10 1 Bottletop Dispenser, without Recirculation Valve; 1 to 10 mL
₹25,100.00INR / Each
COMPARE
30 2.5 Bottletop Dispenser, without Recirculation Valve; 2.5 to 30 mL
₹37,600.00INR / Each
COMPARE
60 5 Bottletop Dispenser, without Recirculation Valve; 5 to 60 mL
₹37,600.00INR / Each
COMPARE
100 10 Bottletop Dispenser, without Recirculation Valve; 10 to 100 mL
₹45,300.00INR / Each