Cole-Parmer Bottletop Dispenser with Calibration; 2.5 to 30 mL

Representative image only.

Cole-Parmer Bottletop Dispenser with Calibration; 2.5 to 30 mL

Cole-Parmer | Item: 1597774

Clean, safe and precise dispensing at very affordable prices

₹37,600.00INR / Each

₹37,600.00INR / Each
Product Description

Includes

five adapters, telescopic tube, calibration tool, and calibration certificate supplied by the manufacturer.

Key Features

  • Set and lock volume for optimized accurate results
  • Available in six volume ranges from 0.25 mL to 100 mL
  • Fully autoclavable and superior chemical compatibility
  • Jam resistant with smooth plunger movement
  • 1 year warranty

More About this Item

Cole-Parmer’s bottletop dispenser offers great performance over long period of time. Spring-less technology enables customer to use both organic and inorganic reagents. Jam-free piston not only helps for free piston movement but also ensures service-free operation. For flexibility of work the adapter is designed to rotate 360° which are available in various sizes - 28mm, 32mm, 36mm, 40mm and 45mm. The PTFE piston with ETP O-ring provide frictionless movement reducing the occurrence of bubbles in your dispenses.

1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Min Dispense Volume (mL)2.5
  • Max Dispense Volume (mL)30
  • Division (mL)0.2
  • Accuracy±0.6%
  • Reproducibility±0.2%
  • AutoclavableYes
  • Wetted PartsPTFE, ETP,borosilicate glass, PP, PFA,and FEP
  • DescriptionBottletop Dispenser, without Recirculation Valve; 2.5 to 30 mL
  • Warranty1 year
Reviews
Q&A