Cole-Parmer® Bottle Top Dispensers with Plastic Bottle
Excellent chemical compatibility and precise dispensing
  • Easy to use with simple, quick volume adjustments
Dispenser is complete with plastic bottle and unique PTFE piston with intergral o-ring provides smooth motion and high accuracy. Features a built-in 30-mm threaded screw top and include adapters for 28-, 32-, 36-, 40- and 45-mm bottles. Dispenser is easy to calibrate and disassemble for cleaning.
70,200.00 - ₹85,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
COMPARE
0.2 10 1
₹70,200.00INR / Each
COMPARE
0.5 30 2.5
₹84,200.00INR / Each
COMPARE
1 60 5
₹85,200.00INR / Each