Cole-Parmer® Bottle Top Dispensers with Glass Bottle
Unique PTFE piston with intergral o-ring provides smooth motion and high accuracy. Features a built-in 30-mm threaded screw top and include adapters for 28-, 32-, 36-, 40- and 45-mm bottles. Dispenser is easy to calibrate and disassemble for cleaning.
73,600.00 - ₹103,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
COMPARE
0.05 2.5 0.25
₹73,600.00INR / Each
COMPARE
0.1 5 0.5
₹86,900.00INR / Each
COMPARE
0.2 10 1
₹74,100.00INR / Each
COMPARE
0.5 30 2.5
₹88,700.00INR / Each
COMPARE
1 60 5
₹103,000.00INR / Each
COMPARE
1 60 5
₹93,800.00INR / Each