Cole-Parmer Bioprocess Storage Containers
Alternative for stainless steel drums that address the concerns of storing 3-D single-use bags
  • Designed for fluid and nonfluid applications, containers are foldable to save space in shipment
  • Smooth surface inhibits bacterial growth
  • Versatile functionality—containers are collapsible and stackable
  • Offered for storage and shipment, ranging from 50- to 1000-L capacities
Cleanroom Bioprocess Storage Containers enhance storage capability within controlled environments. These hygienically designed containers are foldable for efficient storage and offer great accessibility via open access to drain tubes.
STORAGE RANGE [CLEANROOM]
Enhances storage capability within controlled environments
Foldable for storage
Great accessibility for cleaning
Enhances your sustainability
195,000.00 - ₹552,000.00INR / Pkg
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Bioprocess Storage Container, 50 L; 1/Ea
₹195,000.00INR / Pkg
COMPARE
Bioprocess Cleanroom Storage Container, 250 L; 1/Ea
₹208,000.00INR / Pkg
COMPARE
Bioprocess Cleanroom Storage Container, 500 L; 1/Ea
₹475,000.00INR / Pkg
COMPARE
Bioprocess Cleanroom Storage Container, 600 L; 1/Ea
₹533,000.00INR / Pkg
COMPARE
Bioprocess Cleanroom Storage Container, 1000 L; 1/Ea
₹552,000.00INR / Pkg