Cole-Parmer Bioprocess Cleanroom Storage Container, 600 L; 1/Ea
Representative image only.
Cole-Parmer Bioprocess Cleanroom Storage Container, 600 L; 1/Ea
Cole-Parmer | Item: 1595928
Alternative for stainless steel drums that address the concerns of storing 3-D single-use bags
₹533,000.00INR / Pkg
₹533,000.00INR / Pkg
Product Description
Key Features
  • Designed for fluid and nonfluid applications, containers are foldable to save space in shipment
  • Smooth surface inhibits bacterial growth
  • Versatile functionality—containers are collapsible and stackable
  • Offered for storage and shipment, ranging from 50- to 1000-L capacities
More About this Item
Cleanroom Bioprocess Storage Containers enhance storage capability within controlled environments. These hygienically designed containers are foldable for efficient storage and offer great accessibility via open access to drain tubes.
STORAGE RANGE [CLEANROOM]
Enhances storage capability within controlled environments
Foldable for storage
Great accessibility for cleaning
Enhances your sustainability
Specifications
  • DescriptionBioprocess Cleanroom Storage Container, 600 L; 1/Ea