Cole-Parmer® Balance Draft Shield
Clear acrylic prevents drafts from affecting weighings
  • No assembly required
  • Mouse hole in back panel allows for easy access of power cords
  • Minimizes air movement around your balance
Minimize the air flow around your balance for faster, more stable reading with the clear acrylic draft shield. Easy to install over your balance. Raised edges on top of enclosure allow secure placement of frequently used weighing accessories.
26,600.00 - ₹44,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Draft Shield Height (in) Draft Shield Width (in) Length (")
Availability Pricing
COMPARE
14 3/4 13 13 1/2
₹26,600.00INR / Each
COMPARE
16 3/4 15 15 1/2
₹31,600.00INR / Each
COMPARE
19 3/4 18 18 1/2
₹44,000.00INR / Each