Cole-Parmer® Aspirating Serological Pipettes
Provides rapid fluid dispensing
  • Made from transparent medical grade polystyrene (PS) sterilized by gamma irradiation
  • Tested for endotoxins, cytotoxicity, and are guaranteed nonhemolytic
  • Certified DNase-, RNase-free, and nonpyrogenic
Plastic serological pipettes are supplied without graduations and filter plugs for vacuum aspirating and free-flowing applications. They are ideal for tissue culture applications when aspirating larger volumes from dishes, flasks, or roller bottles. Individually wrapped.
12,100.00 - ₹19,000.00INR / Case of 200
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL)
Availability Pricing
COMPARE
2
₹19,000.00INR / Case of 500
COMPARE
5
₹16,200.00INR / Case of 200
COMPARE
10
₹12,100.00INR / Case of 200