Cole-Parmer Sterile Aspirating Serological Pipettes, 10 mL, Individually Wrap; 200/Cs
Representative image only.
Cole-Parmer Sterile Aspirating Serological Pipettes, 10 mL, Individually Wrap; 200/Cs
Cole-Parmer | Item: 0439510
Provides rapid fluid dispensing
₹12,100.00INR / Case of 200
₹12,100.00INR / Case of 200
Buy It Together
Current Item
Cole-Parmer Sterile Aspirating Serological Pipettes,  10 mL, Individually Wrap; 200/Cs
Cole-Parmer Sterile Aspirating Serological Pipettes, 10 mL, Individually Wrap; 200/Cs
0439510
₹12,100.00INR / Case of 200
+ + =
Total Price 51300.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Made from transparent medical grade polystyrene (PS) sterilized by gamma irradiation
  • Tested for endotoxins, cytotoxicity, and are guaranteed nonhemolytic
  • Certified DNase-, RNase-free, and nonpyrogenic
More About this Item
Plastic serological pipettes are supplied without graduations and filter plugs for vacuum aspirating and free-flowing applications. They are ideal for tissue culture applications when aspirating larger volumes from dishes, flasks, or roller bottles. Individually wrapped.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • SterileYes
  • Max Volume (mL)10
  • Overall Length (InchShort)12
  • Body MaterialPolystyrene
  • Color CodeTransparent
  • DescriptionSterile Aspirating Serological Pipettes, 10 mL, Individually Wrap; 200/Cs