Cole-Parmer® API ASTM Plain Form Hydrometers
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Conform to ASTM Specifications
  • Metal Ballast
  • Individually Serialized
These hydrometers conform to the accuracy requirements for ASTM hydrometers, and have their scales affixed using a clear eposy for increased durability. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
10,500.00 - ₹16,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
9/21 Degree API ASTM 2H Hydrometer
₹15,900.00INR / Each
COMPARE
19/31 Degree API ASTM 3H Hydrometer
₹10,500.00INR / Each
COMPARE
39/51 Degree API ASTM 5H Hydrometer
₹16,600.00INR / Each
COMPARE
49/61 Degree API ASTM 6H Hydrometer
₹16,300.00INR / Each