Cole-Parmer® Ammonia Ion-Selective Electrodes
Ideal for high-purity water, fish tanks, sea water, waste water, plating baths, air/stack gases, and biological samples
  • Reference electrode and half-cell combined in one housing
  • Measure concentrations up to 17,000 ppm
  • Available with loose membranes or pre-membraned caps depending on the user's skill and comfort with membrane installation
Easy-to-use, single-junction electrodes feature durable, epoxy bodies. Electrode design minimizes errors and provides accurate results within 30 seconds, depending on solution concentration. Concentration range is 0.01 to 17,000 ppm. Use over a 0 to 50°C temperature range and above pH 11. BNC connector and one-meter cable allow you to use with almost any existing meter.
60,000.00 - ₹76,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
ISE Electrode, Ammonia, Loose Membrane Cap; BNC
₹60,000.00INR / Each
COMPARE
ISE Electrode, Ammonia, Pre-Membraned Cap; BNC
₹76,400.00INR / Each