Cole-Parmer ISE Electrode, Ammonia, Pre-Membraned Cap; BNC
Representative image only.
Cole-Parmer ISE Electrode, Ammonia, Pre-Membraned Cap; BNC
Ideal for high-purity water, fish tanks, sea water, waste water, plating baths, air/stack gases, and biological samples
₹86,100.00INR / Each
₹86,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer ISE Electrode, Ammonia, Pre-Membraned Cap; BNC
Cole-Parmer ISE Electrode, Ammonia, Pre-Membraned Cap; BNC
2751200
₹86,100.00INR / Each
+ + +
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
₹64,600.00INR / Each
+ =
Total Price 302300.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
15-mL of electrolyte, replacement membranes or membrane caps, filling pipette, and instructions. Order standards, ionic strength adjustors, and fill solution separately.
Key Features
  • Reference electrode and half-cell combined in one housing
  • Measure concentrations up to 17,000 ppm
  • Available with loose membranes or pre-membraned caps depending on the user's skill and comfort with membrane installation
More About this Item
Easy-to-use, single-junction electrodes feature durable, epoxy bodies. Electrode design minimizes errors and provides accurate results within 30 seconds, depending on solution concentration. Concentration range is 0.01 to 17,000 ppm. Use over a 0 to 50°C temperature range and above pH 11. BNC connector and one-meter cable allow you to use with almost any existing meter.
Specifications
  • Electrode TypeSingle junction, gas sensing
  • Electrode Body MaterialEpoxy
  • Ion-Selective ElectrodeAmmonia
  • Min pH (pH)11
  • Max pH (pH)14
  • Max ISE (ppm)17000
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • ConnectorBNC
  • Diameter (mm)12
  • Cable Length (m)1
  • DescriptionISE Electrode, Ammonia, Pre-Membraned Cap; BNC