Cole-Parmer® Amber Narrow-Mouth Plastic Bottles, PP
Store chemical samples safely
  • Leakproof polypropylene caps
  • Built-in shoulder loop for easier handling
  • Autoclavable
Use these sturdy bottles whenever excellent chemical resistance is desired for most acids, bases, and alcohols. Protects light-sensitve liquids by reducing UV light transmissions. Ideal for storing chemicals and standards, as well as for collecting and storing distilled water. Larger bottles feature built-in shoulder loop.
4,800.00 - ₹6,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
2 38-430 Polypropylene(PP)
₹4,800.00INR / Each
COMPARE
4 38-430 Polypropylene(PP)
₹6,700.00INR / Each