Cole-Parmer® Adjustable-Angle Industrial Bimetal Thermometers
NEMA 4X/IP66 protection with all-welded stainless steel construction for the toughest of jobs
  • Hermetically sealed case prevents entry of moisture, interior corrosion and coil freeze up
  • Anti-parallax dial for precise readings of both scales
  • Silicone-dampened coil reduces pointer flutter and improves response time during high vibration applications
  • Adjustable-angle connection can be rotated 360 degrees
  • External adjustment reduces maintenance
  • NEMA 4X/IP66 protection
Use these industrial dual-scale thermometers for a wide variety of applications from petroleum plants to food and beverage. Various temperature ranges and dial sizes can accommodate anyone. Each thermometer can perform quick and limited span adjustments. Dry; no liquid fill.
17,800.00 - ₹21,000.00INR / Each
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Temperature (° C) Min Temperature (° C)
Availability Pricing
COMPARE
3 120 -20
₹17,800.00INR / Each
COMPARE
3 120 -20
₹17,800.00INR / Each
COMPARE
3 120 -20
₹17,800.00INR / Each
COMPARE
3 120 -20
₹17,800.00INR / Each
COMPARE
3 120 -20
₹17,800.00INR / Each
COMPARE
3 50 -40
₹17,800.00INR / Each
COMPARE
3 50 -40
₹17,800.00INR / Each
COMPARE
3 50 -40
₹17,800.00INR / Each
COMPARE
3 50 -40
₹17,800.00INR / Each
COMPARE
3 50 -40
₹17,800.00INR / Each
COMPARE
3 290 10
₹17,800.00INR / Each
COMPARE
3 290 10
₹17,800.00INR / Each
COMPARE
3 290 10
₹17,800.00INR / Each
COMPARE
3 290 10
₹17,800.00INR / Each
COMPARE
3 290 10
₹17,800.00INR / Each
COMPARE
5 120 -20
₹21,000.00INR / Each
COMPARE
5 120 -20
₹21,000.00INR / Each
COMPARE
5 120 -20
₹21,000.00INR / Each
COMPARE
5 120 -20
₹21,000.00INR / Each
COMPARE
5 120 -20
₹21,000.00INR / Each
Showing 1 - 20 of 30