Cole-Parmer® 12-mm High-Accuracy, Single-Junction, pH Electrodes
Economical and efficient combination—buy a protective housing assembly, then simply replace the electrode as needed
  • Use for many applications in a variety of housings
High-accuracy electrodes are perfect for testing where temperature variations make many pH electrodes unreliable. These single-junction electrodes feature an innovative design that stabilizes readings in less than 20 seconds, with an accuracy of ±0.02 pH! The pH value stays constant at any given temperature up to pH 13. Maximum pressure is 100 psi at 25°>C.
29,300.00 - ₹53,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
High-Accuracy pH Electrode, 10-ft Cable
₹29,300.00INR / Each
COMPARE
High-Accuracy pH Electrode, 1 k Ohms Pt RTD, 10-ft Cable
₹33,900.00INR / Each
COMPARE
High-Accuracy pH Electrode, 100 Ohms Pt RTD, 25-ft Cable
₹53,700.00INR / Each