Cole-Parmer® 12-mm General-Purpose, Single-Junction pH Electrodes
Save money—just replace electrode
  • Purchase a separate housing and simply replace electrodes as needed
  • Dome bulb offers better protection from breakage
  • Stable in low-ionic strength solutions
  • Offers resistance to sodium ion error
  • Compatible with most systems
General-purpose single-junction electrodes are economical yet accurate. Can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°>C.
23,300.00 - ₹30,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Probe, SJ/Glass/100Ohm RTD/12 mm/10'; Tinned/BNC
₹29,500.00INR / Each
COMPARE
General Purpose pH Electrode, 10-ft Cable
₹24,400.00INR / Each
COMPARE
General Purpose pH Electrode, 25-ft Cable
₹23,300.00INR / Each
COMPARE
General Purpose pH Electrode, 100 Ohms Pt RTD, 25-ft Cable
₹30,700.00INR / Each