Cole-Parmer® 12-mm Diameter ORP Electrodes
Use for many applications in a variety of industrial housings
  • Purchase a separate housing and simply replace electrodes as needed
  • Dome bulb offers better protection from breakage
  • Stable in low-ionic strength solutions
  • Offers resistance to sodium ion error
  • Compatible with most systems
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212°>F (-5 to 100°>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230°>F (-5 to 110°>C).
35,200.00 - ₹49,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Single-Junction ORP Electrode, Platinum Sensor, 10-ft Cable 05994-21
₹35,200.00INR / Each
COMPARE
Single-Junction ORP Electrode, Gold Sensor, 10-ft Cable
₹39,300.00INR / Each
COMPARE
ORP electrode, double-junction, platinum band, 10-ft cable
₹39,400.00INR / Each
COMPARE
Double-Junction ORP Electrode, Gold Sensor, 10-ft Cable
₹49,900.00INR / Each