Cole-Parmer Double-Junction ORP Electrode, Gold Sensor, 10-ft Cable
Representative image only.
Cole-Parmer Double-Junction ORP Electrode, Gold Sensor, 10-ft Cable
Cole-Parmer | Item: 2700625
Use for many applications in a variety of industrial housings
₹49,900.00INR / Each
₹49,900.00INR / Each
Product Description
Includes
All models include a cable with BNC connections. Order industrial housings seperately.
Key Features
  • Purchase a separate housing and simply replace electrodes as needed
  • Dome bulb offers better protection from breakage
  • Stable in low-ionic strength solutions
  • Offers resistance to sodium ion error
  • Compatible with most systems
  • 1 year warranty
More About this Item
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212°>F (-5 to 100°>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230°>F (-5 to 110°>C).
1 Year Warranty
Specifications
  • ApplicationsWater/wastewater, plating baths
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • Max Pressure (PSI)100
  • ConnectorBNC
  • Diameter (mm)12
  • Cable Length (m)3
  • DescriptionDouble-Junction ORP Electrode, Gold Sensor, 10-ft Cable