Cole-Parmer® 10-32 Thread to Hose Barb Fittings, Straight Adapter, Natural Polycarbonate
Easily transition from threaded connection to soft tubing
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, DEHP free, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Threaded barb adapters provide a quick way to transition from soft tubing to a system that accepts a threaded connection. Threads provide a tight and secure connection; a specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
Select from adapters with 10-32 UNF(M) or 10-32 UNF Taper(M) thread. The 10-32 Taper(M) fittings have a tapered 10-32 thread which can create a better seal without the use of additional thread sealants or O-rings. The tapered design creates more compression and a tighter radial seal than the standard, straight UNF threads.
1,200.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Thread size
Availability Pricing
COMPARE
1/16 10-32 Taper(M)
₹1,200.00INR / Pkg of 10
COMPARE
1/16 10-32 UNF(M)
₹1,200.00INR / Pkg of 10
COMPARE
3/32 10-32 Taper(M)
₹1,200.00INR / Pkg of 10
COMPARE
3/32 10-32 UNF(M)
₹1,200.00INR / Pkg of 10
COMPARE
1/8 10-32 Taper(M)
₹1,200.00INR / Pkg of 10
COMPARE
1/8 10-32 UNF(M)
₹1,200.00INR / Pkg of 10
COMPARE
5/32 10-32 Taper(M)
₹1,200.00INR / Pkg of 10
COMPARE
5/32 10-32 UNF(M)
₹1,200.00INR / Pkg of 10
Showing 1- 8 of 8