Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, Natural Polycarbonate, 10-32 Taper(M) x 1/8" ID; 10/Pk
Representative image only.
Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, Natural Polycarbonate, 10-32 Taper(M) x 1/8" ID; 10/Pk
Cole-Parmer | Item: 5014005
Easily transition from threaded connection to soft tubing
₹1,200.00INR / Pkg of 10
₹1,200.00INR / Pkg of 10
Product Description
Key Features
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, DEHP free, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • 10-32 Taper(M) x 1/8" hose barb
  • Provides more compression and a tighter radial seal than the standard, straight UNF threads
  • Works with a female 10-32 UNF port
  • Manufacturer defect warranty
More About this Item
Threaded barb adapters provide a quick way to transition from soft tubing to a system that accepts a threaded connection. Threads provide a tight and secure connection; a specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
Select from adapters with 10-32 UNF(M) or 10-32 UNF Taper(M) thread. The 10-32 Taper(M) fittings have a tapered 10-32 thread which can create a better seal without the use of additional thread sealants or O-rings. The tapered design creates more compression and a tighter radial seal than the standard, straight UNF threads.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
ISO 9001:2008 Certified Supplier
Specifications
  • Fitting MaterialPolycarbonate
  • Sterilization MethodEtO or gamma irradiation
  • Connection TypeThreaded Taper(M) x Hose Barb
  • Number Of Ports2
  • Thread size10-32 Taper(M)
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Flow PatternStraight
  • ColorNatural
  • Max Temperature (° F)797
  • Max Temperature (° C)425
  • Regulatory ApprovalsNSF-51;RoHS;FDA;Non-DEHP
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, Natural Polycarbonate, 10-32 Taper(M) x 1/8" Hose Barb; 10/Pk
  • WarrantyManufacturer defect