ChromAir Gas Monitoring Badges
Six color-change levels to better isolate exposure readings
  • Ideal for both personal and area monitoring
Gas Monitoring Badges are lightweight colorimetric — ideal for both personal and area monitoring. Designed to accurately provide immediate results while minimizing the effects of humidity and air velocity. Each ChromAir badge shows exposure levels. The scale printed on the badges is based on exposure dose. Badges attach easily to shirt or lab coat collars — order badge clips separately
57,000.00 - ₹66,200.00INR / Box of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
Morphix 380003-10 Gas Monitoring Badges; Ammonia, 10/Box 8656321 Mfr # 380003-10
COMPARE
Ammonia
₹57,000.00INR / Box of 10
Morphix 380008-10 Gas Monitoring Badges; Carbon monoxide, 10/Box 8656322 Mfr # 380008-10
COMPARE
CO
₹57,000.00INR / Box of 10
Morphix 380004-10 Gas Monitoring Badges; Chlorine, 10/Box 8656323 Mfr # 380004-10
COMPARE
Chlorine
₹57,000.00INR / Box of 10
Morphix 380007-10 Gas Monitoring Badges; Formaldehyde, 10/Box 8656324 Mfr # 380007-10
COMPARE
Formaldehyde
₹57,000.00INR / Box of 10
Morphix 380017-10 Gas Monitoring Badges; Glutaraldehyde, 10/Box 8656325 Mfr # 380017-10
COMPARE
Glutaraldehyde
₹66,200.00INR / Box of 10
Morphix 380018-10 Gas Monitoring Badges; Mercury, 10/Box 8656327 Mfr # 380018-10
COMPARE
Mercury