Morphix 380017-10 Gas Monitoring Badges; Glutaraldehyde, 10/Box
Representative image only.
Morphix 380017-10 Gas Monitoring Badges; Glutaraldehyde, 10/Box
Morphix | Mfr: 380017-10| Item: 8656325
Six color-change levels to better isolate exposure readings
₹66,200.00INR / Box of 10
₹66,200.00INR / Box of 10
Product Description
Key Features
  • Ideal for both personal and area monitoring
  • Five exposure levels covering STEL (15 minutes)
  • Minimum detectable concentration: 0.04 to 0.95 ppm
  • Relative Humidity Range: 25 to 85%
More About this Item
Gas Monitoring Badges are lightweight colorimetric — ideal for both personal and area monitoring. Designed to accurately provide immediate results while minimizing the effects of humidity and air velocity. Each ChromAir badge shows exposure levels. The scale printed on the badges is based on exposure dose. Badges attach easily to shirt or lab coat collars — order badge clips separately
Specifications
  • Gases MeasuredGlutaraldehyde
  • SensitivityCross Sensitivity: None known
  • DescriptionGas Monitoring Badges; Glutaraldehyde, 10/Box