Chemical duty diaphragm pumps
Ideal for rotary evaporator
Diaphragm pumps are chemically resistant making it ideal for various applications involving aggressive solvents and acidic vapors. They are an excellent replacement for rotary vane pumps and water aspirators, especially in rough vacuum applications where corrosive vapors are encountered.
12,600.00 - ₹377,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg) Max Vacuum (torr)
Availability Pricing
Ilmvac Diaphragm Vacuum Pump for Rotary Evaporator 1594393 Mfr # MPC 105 T
COMPARE
₹377,000.00INR / Each
Ilmvac Vacuum Hose, 8X18mm, 1M 1594394 Mfr # 828310-4
COMPARE
₹12,600.00INR / Each