Ilmvac Diaphragm Vacuum Pump for Rotary Evaporator
Representative image only.
Ilmvac Diaphragm Vacuum Pump for Rotary Evaporator
Ilmvac | Mfr: MPC 105 T| Item: 1594393
Ideal for rotary evaporator
₹377,000.00INR / Each
₹377,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Diaphragm pumps are chemically resistant making it ideal for various applications involving aggressive solvents and acidic vapors. They are an excellent replacement for rotary vane pumps and water aspirators, especially in rough vacuum applications where corrosive vapors are encountered.
1 Year Warranty
Specifications
  • DescriptionDiaphragm Vacuum Pump for Rotary Evaporator