ChemExpress Personal Organic Vapor Badge
Analysis performed in an AIHA-accredited lab
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
Chem Express™ Personal OV Monitor Badge is a versatile organic vapor monitor which requires no assembly and utilizes a new, flexible, inert charcoal wafer.
Personal monitoring badges are the most cost effective and efficient ways to monitor chemicals in your environment. Badges are easy to use and require minimal training and experience. They are small and light weight and have minimal effect on worker productivity. Badges are diffusive devices that require no additional equipment such as sampling pumps, calibration materials, or chargers. These long term badges have a low sampling rate preventing the exceedance of their capacity. Analysis on all badges is preformed in an AIHA accredited lab.
Uses: 8 hour TWA, 15 minute STEL, 24 hour IAQ
78,100.00 - ₹117,000.00INR / Pkg of 5
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
Chem Express X566-1 OV Badge with Pre Paid Analysis for 1 Organic Vapor Pkg/5 8637809 Mfr # X566-1
COMPARE
One Organic vapor
₹78,100.00INR / Pkg of 5
Chem Express X566-4 OV Badge with Pre Paid Analysis for 4 Organic Vapors Pkg/5 8637810 Mfr # X566-4
COMPARE
Four Organic vapors
₹117,000.00INR / Pkg of 5