Chem Express X566-4 OV Badge with Pre Paid Analysis for 4 Organic Vapors Pkg/5
Representative image only.
Chem Express X566-4 OV Badge with Pre Paid Analysis for 4 Organic Vapors Pkg/5
Chem Express | Mfr: X566-4| Item: 8637810
Analysis performed in an AIHA-accredited lab
₹117,000.00INR / Pkg of 5
₹117,000.00INR / Pkg of 5
Product Description
Key Features
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
  • 1 year warranty
More About this Item
Chem Express™ Personal OV Monitor Badge is a versatile organic vapor monitor which requires no assembly and utilizes a new, flexible, inert charcoal wafer.
Personal monitoring badges are the most cost effective and efficient ways to monitor chemicals in your environment. Badges are easy to use and require minimal training and experience. They are small and light weight and have minimal effect on worker productivity. Badges are diffusive devices that require no additional equipment such as sampling pumps, calibration materials, or chargers. These long term badges have a low sampling rate preventing the exceedance of their capacity. Analysis on all badges is preformed in an AIHA accredited lab.
Uses: 8 hour TWA, 15 minute STEL, 24 hour IAQ
1 Year Warranty
Specifications
  • Gases MeasuredFour Organic vapors
  • DescriptionOV Badge with Pre Paid Analysis for 4 Organic Vapors Pkg/5