CELLTREAT® Plasteur® Pasteur Pipettes
Safety and convenience of plastic
  • Help prevent glove and skin punctures with flexible plastic—more durable than glass pipettes
  • Efficient—plastic won't break in transit so you get to use all the pipettes in a box
  • Longer 9" pipettes make it easier when working with larger vessels
  • Choose from virgin polystyrene or chemical-resistant polypropylene, bulk or individually wrapped packing
  • Polypropylene pipettes may be autoclaved
16,800.00 - ₹38,400.00INR / Case of 200
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Overall Length (in) Description
Availability Pricing
CELLTREAT Scientific Products 229270 Plasteur Pasteur Pipette, PS, 5.75" L, Ind Wrapped, Sterile; 200/cs WW-34567-80 Mfr # 229270
COMPARE
Polystyrene 5.75 Plasteur Pasteur Pipette, PS, 5.75\" L, Ind Wrapped, Sterile; 200/cs
₹16,800.00INR / Case of 200
CELLTREAT Scientific Products 229276 Plasteur Pasteur Pipette, PP, 5.75" L, Bulk Packed, Sterile; 500/cs WW-34567-82 Mfr # 229276
COMPARE
Polypropylene 5.75 Plasteur Pasteur Pipette, PP, 5.75\" L, Bulk Packed, Sterile; 500/cs
₹26,100.00INR / Case of 500
CELLTREAT Scientific Products 229277 Plasteur Pasteur Pipette, PP, 5.75" L, Bulk Packed, Nonsterile; 500/cs WW-34567-83 Mfr # 229277
COMPARE
Polypropylene 5.75 Plasteur Pasteur Pipette, PP, 5.75\" L, Bulk Packed, Nonsterile; 500/cs
₹20,600.00INR / Case of 500
CELLTREAT Scientific Products 229280 Plasteur Pasteur Pipette, PS, 9" L, Ind Wrapped, Sterile; 200/cs WW-34567-85 Mfr # 229280
COMPARE
Polystyrene 9 Plasteur Pasteur Pipette, PS, 9\" L, Ind Wrapped, Sterile; 200/cs
₹20,300.00INR / Case of 200
CELLTREAT Scientific Products 229281 Plasteur Pasteur Pipette, PP, 9" L, Ind Wrapped, Sterile; 200/cs WW-34567-86 Mfr # 229281
COMPARE
Polypropylene 9 Plasteur Pasteur Pipette, PP, 9\" L, Ind Wrapped, Sterile; 200/cs
₹23,100.00INR / Case of 200
CELLTREAT Scientific Products 229285 Plasteur Pasteur Pipette, PS, 9" L, Bulk Packed, Sterile; 500/cs WW-34567-87 Mfr # 229285
COMPARE
Polystyrene 9 Plasteur Pasteur Pipette, PS, 9\" L, Bulk Packed, Sterile; 500/cs
₹38,400.00INR / Case of 500
CELLTREAT Scientific Products 229286 Plasteur Pasteur Pipette, PP, 9" L, Bulk Packed, Sterile; 500/cs WW-34567-88 Mfr # 229286
COMPARE
Polypropylene 9 Plasteur Pasteur Pipette, PP, 9\" L, Bulk Packed, Sterile; 500/cs
₹27,100.00INR / Case of 500
CELLTREAT Scientific Products 229287 Plasteur Pasteur Pipette, PP, 9" L, Bulk Packed, Nonsterile; 500/cs WW-34567-89 Mfr # 229287
COMPARE
Polypropylene 9 Plasteur Pasteur Pipette, PP, 9\" L, Bulk Packed, Nonsterile; 500/cs
₹20,600.00INR / Case of 500
Showing 1- 8 of 8