CELLTREAT® Electronic Pipet Controller Accessories
Replacement parts for your CELLTREAT® Electronic Pipette Controller
  • Adjust speed with the turn of a dial
  • Obtain up to 2500 pipettings (up to 8 hours of use) on a single charge
This CellTreat pipette controller adapts to fill and dispense 1- to 100-mL sized pipettes. Control speed with the turn of a dial, and direction with low-pressure buttons. Avoid accidental over aspiration, contamination, and damage to the controller from volatile vapors with the included filter.
3,400.00 - ₹13,900.00INR / Case
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
CELLTREAT Scientific Products 230202 AC Power Adapter and Charger for Electronic Pipet Controller; 120 VAC WW-25200-78 Mfr # 230202
COMPARE
AC Power Adapter and Charger for Electronic Pipet Controller; 120 VAC
₹13,900.00INR / Case
CELLTREAT Scientific Products 230205 Rubber Grommet for Electronic Pipet Controller; 1/Ea WW-25200-81 Mfr # 230205
COMPARE
Rubber Grommet for Electronic Pipet Controller; 1/Ea
₹10,000.00INR / Case of 1
CELLTREAT Scientific Products 230206 Filter for Electronic Pipet Controller, 0.45 µm; 2/Cs WW-25200-83 Mfr # 230206
COMPARE
Filter for Electronic Pipet Controller, 0.45 µm; 2/Cs
₹3,400.00INR / Case of 2