CELLTREAT Scientific Products 230206 Filter for Electronic Pipet Controller, 0.45 µm; 2/Cs
Representative image only.
CELLTREAT Scientific Products 230206 Filter for Electronic Pipet Controller, 0.45 µm; 2/Cs
CELLTREAT Scientific Products | Mfr: 230206| Item: 2520083
Replacement parts for your CELLTREAT® Electronic Pipette Controller
₹8,300.00INR / Case of 2
₹8,300.00INR / Case of 2
Product Description
Includes
adhesive mounting bracket, rubber grommet/nosepiece, 0.45-µm pore filter, rechargeable battery, and 120 VAC power adapter/charger.
Key Features
  • Adjust speed with the turn of a dial
  • Obtain up to 2500 pipettings (up to 8 hours of use) on a single charge
More About this Item
This CellTreat pipette controller adapts to fill and dispense 1- to 100-mL sized pipettes. Control speed with the turn of a dial, and direction with low-pressure buttons. Avoid accidental over aspiration, contamination, and damage to the controller from volatile vapors with the included filter.
Specifications
  • Compatible WithCELLTREAT® Electronic Pipet Controller
  • DescriptionFilter for Electronic Pipet Controller, 0.45 µm; 2/Cs