CellSTACK Culture Chambers
Ideal for intermediate to large scale culture processes
  • Innovative stacking design lets you achieve 25,440 cm2 cell growth area
  • Tissue culture (TC) treated for even cell attachment
With Corning® CellSTACK culture chambers, achieving even dispersion of cell media for optimal growth rates in a small footprint has never been easier! Two 26-mm diameter filling and venting ports allow direct access to the chamber bottom, providing greater flexibility for sterile filling and emptying by pouring, pipetting, or via tubing in a fully enclosed system. The standard 33-mm threaded caps have 0.2-µm pore, hydrophobic membranes sealed directly to the caps, allowing gas exchange while minimizing risk of contamination.
Manufactured from USP Class VI polystyrene for excellent optical clarity and mechanical strength, CellSTACK is available in five sizes to meet your needs to scale up your cell culture. All culture chambers are sterilized by gamma irradiation and certified nonpyrogenic.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Material Width (in)
Availability Pricing
Corning 3268 Polystyrene CellSTACK - 1 Chamber with Vent Caps, 8 per Case WW-15941-83 Mfr # 3268
COMPARE
Call For Price
Corning 3269 Polystyrene CellSTACK - 2 Chamber with Vent Caps, 5 per Case WW-15941-84 Mfr # 3269
COMPARE
Call For Price
Corning 3319 Polystyrene CellSTACK - 5 Chamber with Vent Caps, 2 per Case WW-15941-85 Mfr # 3319
COMPARE
Call For Price
Corning 3270 Polystyrene CellSTACK - 10 Chamber with Vent Caps, 2 per Case WW-15941-86 Mfr # 3270
COMPARE
Call For Price
Corning 3272 Polystyrene CellSTACK - 40 Chamber with Vent Caps, 2 per Case WW-15941-87 Mfr # 3272
COMPARE
Call For Price
Corning 3331 Stacking Device, polystyrene WW-15941-88 Mfr # 3331
COMPARE
Call For Price
Corning 3969 Solid Cap, non vented WW-15941-90 Mfr # 3969
COMPARE
Call For Price
Corning 3333 Filling Cap with 1/4" (6.4mm) ID Tubing with a Male MPC Coupling and a Female MPC End Cap WW-15941-91 Mfr # 3333
COMPARE
Call For Price
Corning 3328 Filling Cap with a female MPC coupling, 1/4" (6.4mm) ID fitting and a Male MPC Polycarbona WW-15941-92 Mfr # 3328
COMPARE
Call For Price
Corning 3329 Filling Cap with a female MPC coupling, 3/8" (9.5mm) ID fitting and a Male MPC Polycarbona WW-15941-93 Mfr # 3329
COMPARE
Call For Price
Corning 3334 Filling Cap with a male MPC coupling, 1/4" (6.4mm) ID fitting and a female MPC Polycarbona WW-15941-94 Mfr # 3334
COMPARE
Call For Price
Corning 3339 Filling Cap with a male MPC coupling, 3/8" (9.5mm) ID fitting and a female MPC Polycarbona WW-15941-95 Mfr # 3339
COMPARE
Call For Price
Showing 1- 12 of 12