Corning 3334 Filling Cap with a male MPC coupling, 1/4" (6.4mm) ID fitting and a female MPC Polycarbona
Representative image only.
Corning 3334 Filling Cap with a male MPC coupling, 1/4" (6.4mm) ID fitting and a female MPC Polycarbona
Corning | Mfr: 3334| Item: 1594194
Ideal for intermediate to large scale culture processes
Call For Price
Call For Price
Product Description
Includes
standard 33-mm vented caps.
Key Features
  • Innovative stacking design lets you achieve 25,440 cm2 cell growth area
  • Tissue culture (TC) treated for even cell attachment
More About this Item
With Corning® CellSTACK culture chambers, achieving even dispersion of cell media for optimal growth rates in a small footprint has never been easier! Two 26-mm diameter filling and venting ports allow direct access to the chamber bottom, providing greater flexibility for sterile filling and emptying by pouring, pipetting, or via tubing in a fully enclosed system. The standard 33-mm threaded caps have 0.2-µm pore, hydrophobic membranes sealed directly to the caps, allowing gas exchange while minimizing risk of contamination.
Manufactured from USP Class VI polystyrene for excellent optical clarity and mechanical strength, CellSTACK is available in five sizes to meet your needs to scale up your cell culture. All culture chambers are sterilized by gamma irradiation and certified nonpyrogenic.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • DescriptionFilling Cap with a male MPC coupling, 1/4" (6.4mm) ID fitting and a female MPC Polycarbona