CELLspin Cell Culture Spinner Systems
Offers unique oscillating movement that guarantees low shear force while maintaining optimum mixing
  • Fits inside a standard-size incubator
  • Order Spinner Flasks to suit your volume needs—sold separately
CELLspin is a programmable spinning system with remote control for stirring culture volumes from 50 to 1000 mL at up to 75 rpm. The unit duplicates gentle stirring conditions in up to four spin flasks. Each flask is fitted with specially rounded glass impellers that provide oscillating circular movement to guarantee low shear force while maintaining optimum mixing.
The pendular angle and interval between turns of the impellers can be programmed individually. The CELLspin also performs continuous circular stirring. The remote controller has a digital display and advanced features such as cyclical mode, ramp up, down, forward, and reverse.
The complete unit includes a timer with alarm. The system is CO2 and moisture resistant. Two CELLspin systems fit inside a standard-size incubator. Measures 11-5/8"L x 13"W x 2-3/8"H. The control unit and stirring platform are also sold separately.
412,000.00 - ₹953,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Pfeiffer CELLspin Spinner System; 100 to 240 VAC, 50/60 Hz 1305100 Mfr # 183001
COMPARE
Spinner System; 100 to 240 VAC, 50/60 Hz
₹953,000.00INR / Each
Pfeiffer CELLspin Spinner System Control Unit, with Power Supply; 100 to 240 VAC, 50/60 Hz 2440617 Mfr # 183013
COMPARE
Control Unit, with Power Supply; 100 to 240 VAC, 50/60 Hz
₹412,000.00INR / Each
Pfeiffer CELLspin Spinner System Stirring Platform with 4 Stirring Positions and Control Cable 2440618 Mfr # 183015
COMPARE
Stirring Platform with 4 Stirring Positions and Control Cable
₹438,000.00INR / Each