CDS Analytical Empore™ Organic SPE Disks
Efficient alternative to liquid extraction for sample preparation
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
The Empore organic SPE disks are ideal for solid phase extraction of large water samples. Due to a large surface area, these disks offer better flow rate and higher sample capacity than small volume cartridges. These disks are approved by the US EPA for many organic analytes and are recognized as an alternative test method. They are safe to use, efficient, and provide excellent solvent reduction.
180,000.00 - ₹295,000.00INR / Pkg of 60
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Description
Availability Pricing
CDS Analytical  2215 Empore™ C-18 Disk, 47mm; 60/PK 3521324 Mfr # G2028C
COMPARE
47 CDS Analytical 2215 Empore™ C-18 Disk, 47mm; 60/PK
₹180,000.00INR / Pkg of 60
CDS Analytical  2241 Empore™ SDB-RPS Disk, 47mm; 60/PK 3521328 Mfr # G2048C
COMPARE
47 CDS Analytical 2241 Empore™ SDB-RPS Disk, 47mm; 60/PK
₹295,000.00INR / Pkg of 60