CDS Analytical Empore™ Oil & Grease SPE Disks
Offers fast flow rates and high throughput
  • Reduces solvent volumes and time for eluate evaporation
  • Optimized to meets EPA Method 1664 requirements
The Empore oil and grease disks offer a faster flow rate and less filter cogging than most other SPE filters. The filters are designed with a fluoropolymer matrix embedded with C-18 coated particles placed between two layers of polypropylene prefilter material. The trilaminar construction eliminates the need for costly and messy prefilter materials on most samples.
102,000.00 - ₹179,000.00INR / Pkg of 60
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Description
Availability Pricing
CDS Analytical  2270 Empore™ Oil/Grease Extraction Disk, 47mm; 60/PK 3521309 Mfr # G2073
COMPARE
47 Fluoropolymer matrix Oil/Grease Extraction Disk, 47mm; 60/PK
₹102,000.00INR / Pkg of 60
CDS Analytical  2370 Empore™ Oil/Grease Extraction Disk, 90mm; 30/PK 3521310 Mfr # G2083
COMPARE
90 Fluoropolymer matrix Oil/Grease Extraction Disk, 90mm; 30/PK
₹179,000.00INR / Pkg of 30