Casella Microdust Pro Aerosol Monitoring System
Scrolling graph of real-time data for quick and easy field analysis
  • Capture real-time data with fast warm-up time and update interval
  • Instantly see changes in concentration, even while monitor is still recording
  • Calibrate to any aerosolized particulate, including composite matter, for increased sensitivity
This portable aerosol monitor measures levels of any airborne particulate matter, including dust, smoke, fumes, pollen, and other aerosols produced by many industries. Comfortable, ergonomic housing with detachable probe make this unit ideal for general walk-through surveys. Large graphical LCD features color-coded screens for quick navigation.
Store up to 86,000 data points or 500 discrete measurement sessions in internal memory; at intervals of 1 second to 60 minutes. Each data logging session ends with average and maximum values calculated during the logging period. Included Casella Insight software lets you configure the monitor, download and analyze data, and produce reports.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Casella CEL-712/K1 Aerosol Monitoring System WW-86552-01 Mfr # CEL-712/K1
COMPARE
Aerosol Monitoring System
Call For Price
Casella CEL-712/K1 Dust Detective Kit with sampling pump WW-86552-02 Mfr # 206107A
COMPARE
Dust Detective Kit with sampling pump