Casella CEL-712/K1 Aerosol Monitoring System
Representative image only.
Casella CEL-712/K1 Aerosol Monitoring System
Casella | Mfr: CEL-712/K1| Item: 8655201
Scrolling graph of real-time data for quick and easy field analysis
Call For Price
Call For Price
Product Description
Includes
calibration insert, cleaning bellows, software, USB cable, hard carrying case, and three AA batteries.
Key Features
  • Capture real-time data with fast warm-up time and update interval
  • Instantly see changes in concentration, even while monitor is still recording
  • Calibrate to any aerosolized particulate, including composite matter, for increased sensitivity
  • 2 year warranty
More About this Item
This portable aerosol monitor measures levels of any airborne particulate matter, including dust, smoke, fumes, pollen, and other aerosols produced by many industries. Comfortable, ergonomic housing with detachable probe make this unit ideal for general walk-through surveys. Large graphical LCD features color-coded screens for quick navigation.
Store up to 86,000 data points or 500 discrete measurement sessions in internal memory; at intervals of 1 second to 60 minutes. Each data logging session ends with average and maximum values calculated during the logging period. Included Casella Insight software lets you configure the monitor, download and analyze data, and produce reports.
Two year warrantyConformite Europeenne
Specifications
  • Data LoggingYes
  • Data Storage (samples)15700
  • Logging Intervals2 seconds to 10 minutes
  • Battery3 AA (included)
  • Battery Life20 hours, typical
  • DescriptionAerosol Monitoring System