Burkle Turn ‘n’ Spray Bottles with Overhead Valve
Uninterrupted spraying at any angle, even over your head
  • Adjustable nozzle delivers from a fine mist to powerful stream
  • Parts in contact with media made of PP, PE, silicon and stainless steel
  • Sprays up to 1.2 mL (± 0.1) volume per stroke
The Turn ‘n’ Spray bottles feature a special technology allowing you to spray uninterrupted at any angle or direction from any position. The overhead valve design permits you to reach hard-to-access places easily and without straining. Made of translucent low-density polyethylene (LDPE), the refillable spray bottles can be safely used again and again, saving you money. Wetted materials include LDPE, PP, PE, silicon, and stainless steeel.
3,800.00 - ₹4,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Neck Inside Diameter (ID) (mm) Overall Height (in)
Availability Pricing
Burkle Turn 'n' Spray Spray bottle with trigger sprayer, works at any angle or upside down, 250 ml; 1.2 mL + .1 spray 0604005 Mfr # 0309-0002
COMPARE
250 18 8 45/64
₹3,800.00INR / Each
Burkle Turn 'n' Spray Spray bottle with trigger sprayer, works at any angle or upside down, 500 ml; 1.2 mL + .1 spray 0604006 Mfr # 0309-0005
COMPARE
500 18 9 1/2
₹4,200.00INR / Each
Burkle Turn 'n' Spray Spray bottle with trigger sprayer, works at any angle or upside down, 1000 ml; 1.2 mL + .1 spray 0604007 Mfr # 0309-0010
COMPARE
1000 17.5 11 19/32
₹4,300.00INR / Each