Burkle Trigger Spray Bottles
Suitable for agricultural pesticides and detergents
  • Adjustable nozzle delivers from a fine mist to powerful stream
  • Parts in contact with media made of PP, PE, silicon, and stainless steel
  • Sprays up to 1.2 mL (± 0.1) volume per stroke
These pressureless spray bottles, made of low-density polyethylene (LDPE), feature a long-life trigger-action spray pump allowing them to be successfully used over and over again. The translucent refillable spray bottles are recommended for industrial, laboratory, production, and household use.
2,300.00 - ₹2,600.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Neck Inside Diameter (ID) (mm) Overall Height (in)
Availability Pricing
Burkle Spray bottle with trigger sprayer, 250 ml; 1.2 mL + .1 spray 0604008 Mfr # 0309-1025
COMPARE
250 18 8 45/64
₹2,300.00INR / Each
Burkle Spray bottle with trigger sprayer, 500 ml; 1.2 mL + .1 spray 0604009 Mfr # 0309-1050
COMPARE
500 18 9 1/2
₹2,600.00INR / Each
Burkle Spray bottle with trigger sprayer, 1000 ml; 1.2 mL + .1 spray 0604014 Mfr # 0309-1100
COMPARE
1000 17.5 11 19/32
₹2,600.00INR / Each