Burkle LiquiSampler Valved Liquid Sampler for Open or Closed Containers
Easy, contamination-free sampling
  • Single-handed valve operation makes multilayer and single-point sampling easy
  • Ultra-smooth surface prevents sample, residue, and dirt accumulation
  • Scraping ring marks depth or helps clean outside of sampler when withdrawing
  • Choose chemically-inert PTFE/FEP or economical PP
  • Fits in any container with a minimum opening diameter of 1-1/4" (32 mm)
This valved liquid sampler prevents contamination and is easy to operate. Ultra-pure PTFE/FEP construction is suitable for harsh or critical chemicals, while economical polypropylene (PP) works for aqueous solutions and noncorrosive chemicals. Both material options feature a smooth surface where no crevices or pores exist, eliminating concern for accumulation and cross-contamination. Integral scraping ring can be used to mark depth, or to wipe excess sample off the outside of the tube.
Push-button valve is normally closed. Simply push the button to open the valve and let the sample into the tube. Sampler features threaded connections that are easy to disassemble and clean.
92,000.00 - ₹268,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Burkle 5330-1060 Fluid sampler, PP, gravity feed, vacuum button release, 60 cm x 32 mm; 150 mL 0604171 Mfr # 5330-1060
COMPARE
Valved Drum and Tank Sampler, PP, 60 cm x 32 mm; 150 mL
₹92,000.00INR / Each
Burkle 5330-1100 Fluid sampler, PP, gravity feed, vacuum button release, 100 cm x 32 mm; 250 mL 0604172 Mfr # 5330-1100
COMPARE
Valved Drum and Tank Sampler, PP, 100 cm x 32 mm; 250 mL
₹123,000.00INR / Each
Burkle 5330-1200 Fluid sampler, PP, gravity feed, vacuum button release, 200 cm x 32 mm; 500 mL 0604173 Mfr # 5330-1200
COMPARE
Valved Drum and Tank Sampler, PP, 200 cm x 32 mm; 500 mL
₹183,000.00INR / Each
Burkle 5330-2060 Fluid sampler, PTFE/FEP, gravity feed, vacuum button release, 60 cm x 32 mm; 150 mL 0604174 Mfr # 5330-2060
COMPARE
Valved Drum and Tank Sampler, PTFE/FEP, 60 cm x 32 mm; 150 mL
₹145,000.00INR / Each
Burkle 5330-2100 Fluid sampler, PTFE/FEP, gravity feed, vacuum button release, 100 cm x 32 mm; 250 mL 0604175 Mfr # 5330-2100
COMPARE
Valved Drum and Tank Sampler, PTFE/FEP, 100 cm x 32 mm; 250 mL
₹193,000.00INR / Each
Burkle 5330-2200 Fluid sampler, PTFE/FEP, gravity feed, vacuum button release, 200 cm x 32 mm; 500 mL 0604176 Mfr # 5330-2200
COMPARE
Valved Drum and Tank Sampler, PTFE/FEP, 200 cm x 32 mm; 500 mL
₹268,000.00INR / Each