Burkle Disposable Polystyrene Food-Grade Sampling Spatulas
EU and FDA compliant to safely handle food samples
  • Suitable for food-grade applications
  • Individually packaged in easy-to-open bags
  • Produced and assembled according to cleanroom ISO Class 7
The ergonomically shaped spatulas feature a stable handle and sharp blade allowing it to penetrate directly into paper or plastic sacks—ideal for sampling powders, granulates, and pastes. The spatulas are 10 3/8” (26.3 cm) long with an insertion depth of 6” (15 cm). A transparent sealing sleeve, available on models 06042-40 and 06042-60, securely transports samples and safeguards against possible contamination. Available non-terilized or presterilized via gamma irradiation.
7,700.00 - ₹11,400.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (in) Sterile Description
Availability Pricing
Burkle 5378-0010 Disposable Sampling Spatula with Cover, PS, White, 150 mm; 10/PK 0604240 Mfr # 5378-0010
COMPARE
10 1/2 No Disposable Sampling Spatula with Cover, PS, FDA Compliant, White; 150 mm
₹10,600.00INR / Pkg of 10
Burkle 5378-1009 Disposable Sampling Spatula, PS, White, Sterile, 150 mm; 10/PK 0604259 Mfr # 5378-1009
COMPARE
10 2/5 Yes Disposable Sampling Spatula, PS, FDA Compliant, White, Sterile; 150 mm
₹7,700.00INR / Pkg of 10
Burkle 5378-1010 Disposable Sampling Spatula with Cover PS, White, Sterile; 150 mm; 10/PK 0604260 Mfr # 5378-1010
COMPARE
10 1/2 Yes Disposable Sampling Spatula with Cover PS, FDA Compliant, White, Sterile; 150 mm
₹11,400.00INR / Pkg of 10