Burkle Aluminum Bottles with Tamper-Evident Caps
Easily keep important solutions unaltered
  • Pure aluminum bottle protects light-sensitive chemicals
  • Tamper-evident polypropylene screw cap keeps sample unchanged
  • Perfect for storage and transport of laboratory mixtures as well as food
These rustproof aluminum bottles provide an unbreakable storage solution for important laboratory samples. Bottles can be completely emptied for easy cleaning and sample retrieval. The tamper-evident polypropylene screw cap has an aluminum inner seal. A safety ring is secured when the cap is screwed onto the bottle; opening the bottle breaks the safety ring, indicating signs of interference.
960.00 - ₹2,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Description
Availability Pricing
Burkle 0327-0038 Aluminum Bottle with Tamper-Evident Cap, 38 mL; 1/EA 0604021 Mfr # 0327-0038
COMPARE
38 Aluminum Bottle with Tamper-Evident Cap, 38 mL; 1/EA
₹960.00INR / Each
Burkle 0327-0060 Aluminum Bottle with Tamper-Evident Cap, 60 mL; 1/EA 0604022 Mfr # 0327-0060
COMPARE
60 Aluminum Bottle with Tamper-Evident Cap, 60 mL; 1/EA
₹1,000.00INR / Each
Burkle 0327-0120 Aluminum Bottle with Tamper-Evident Cap, 120 mL; 1/EA 0604023 Mfr # 0327-0120
COMPARE
120 Aluminum Bottle with Tamper-Evident Cap, 120 mL; 1/EA
₹1,300.00INR / Each
Burkle 0327-0300 Aluminum Bottle with Tamper-Evident Cap, 300 mL; 1/EA 0604024 Mfr # 0327-0300
COMPARE
300 Aluminum Bottle with Tamper-Evident Cap, 300 mL; 1/EA
₹1,700.00INR / Each
Burkle 0327-0600 Aluminum Bottle with Tamper-Evident Cap, 600 mL; 1/EA 0604025 Mfr # 0327-0600
COMPARE
600 Aluminum Bottle with Tamper-Evident Cap, 600 mL; 1/EA
₹2,400.00INR / Each
Burkle 0327-1200 Aluminum Bottle with Tamper-Evident Cap, 1200 mL; 1/EA 0604026 Mfr # 0327-1200
COMPARE
1200 Aluminum Bottle with Tamper-Evident Cap, 1200 mL; 1/EA
₹2,800.00INR / Each