BriskHeat® Silicone Rubber Heating Tapes with Adjustable Thermostat Control
Convenient, all-in-one design eliminates the need for a separate controller
Solve a wide range of heating applications without risk of damage due to chemical and moisture spillover. Tapes are available thermostat control. Tapes are made of fine-strand resistance wires that are double insulated with braided fiberglass and knitted into flat tapes; then completely enclosed in highstrength, high-temperature-resistant silicone rubber.
Warning: Heating tapes should not be overlapped. For thermostat control tapes, the sensor
box must make surface contact with the object being heated.
36,100.00 - ₹46,100.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat 051002 Silicone Heating Tape with Adjustable Control, 0.5" W x 2'L, 72W; 120V with Plug 03106-90 0310690 Mfr # THSTAT051002
COMPARE
2 425 120
₹36,100.00INR / Each
BriskHeat 051004 Silicone Heating Tape with Adjustable Control, 0.5" W x 4'L, 144W; 120V with Plug 03106-91 0310691 Mfr # THSTAT051004
COMPARE
4 425 120
₹38,500.00INR / Each
BriskHeat 051006 Silicone Heating Tape with Adjustable Control, 0.5" W x 6'L, 216W; 120V with Plug 0310692 Mfr # THSTAT051008
COMPARE
6 425 120
₹40,100.00INR / Each
BriskHeat 051008 Silicone Heating Tape with Adjustable Control, 0.5" W x 8'L, 288W; 120V with Plug 0310693 Mfr # HSTAT051008
COMPARE
8 425 120
₹38,700.00INR / Each
BriskHeat 051010 Silicone Heating Tape with Adjustable Control, 0.5" W x 10'L, 360W; 120V with Plug 0310694 Mfr # THSTAT051010
COMPARE
10 425 120
₹46,100.00INR / Each
BriskHeat 101002 Silicone Heating Tape with Adjustable Control, 1" W x 2'L, 144W; 120V with Plug 0310695 Mfr # THSTAT101002
COMPARE
2 425 120
₹37,900.00INR / Each
BriskHeat 101004 Silicone Heating Tape with Adjustable Control, 1" W x 4'L, 288W; 120V with Plug 0310696 Mfr # THSTAT101004
COMPARE
4 425 120
₹38,600.00INR / Each
BriskHeat 101006 Silicone Heating Tape with Adjustable Control, 1" W x 6'L, 432W; 120V with Plug 0310697 Mfr # THSTAT101006
COMPARE
6 425 120
₹42,100.00INR / Each
BriskHeat 101008 Silicone Heating Tape with Adjustable Control, 1" W x 8'L, 576W; 120V with Plug 0310698 Mfr # THSTAT101008
COMPARE
8 425 120
₹44,200.00INR / Each
Showing 1- 9 of 9