BriskHeat Adjustable Wrap-Around Tote Tank/IBC Blanket Heater
Avoid freezing and increase flow rates of tote tank/IBC contents
  • Built-in manual reset high-limit thermostat offers added versatility
Provide fast uniform heat to your tote tank/IBC contents with this wrap-around blanket heater. The full coverage plug and play system is constructed of a silicone impregnated cloth facing and liner with a 1/4"-thick fiberglass insulation, and is intended for indoor use. Setup and operation is simple — wrap the blanket around your tote tank, secure it with the adjustable nylon straps, and set your desired temperature. Blanket heater will fit any tote tank width from 40"x 40", to 48" x 48".
87,400.00 - ₹648,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat TOTE-TOP Insulated top cover for adjustable wrap-around tote tank / IBC blanket heaters 0310683 Mfr # TOTE-TOP
COMPARE
₹87,400.00INR / Each
BriskHeat TOTE361-ADJ Tote Tank/Ibc Blanket Heater, 36" , 120 V 0930205 Mfr # TOTE361-ADJ
COMPARE
40 in x 40 in to 48 in x 48 in 160 120
₹537,000.00INR / Each
BriskHeat TOTE362-ADJ Tote Tank /IBC Blanket Heater, 36" , 240V 0930206 Mfr # TOTE362-ADJ
COMPARE
40 in x 40 in to 48 in x 48 in 160 240
₹537,000.00INR / Each
BriskHeat TOTE421-ADJ Tote Tank/Ibc Blanket Heater, 42" , 120 V 0930210 Mfr # TOTE421-ADJ
COMPARE
40 in x 40 in to 48 in x 48 in 160 120
₹600,000.00INR / Each
BriskHeat TOTE481-ADJ Tote Tank/Ibc Blanket Heater, 48" , 120 V 0930215 Mfr # TOTE481-ADJ
COMPARE
40 in x 40 in to 48 in x 48 in 160 120
₹648,000.00INR / Each
BriskHeat TOTE482-ADJ Tote Tank/Ibc Blanket Heater, 48" , 240 V 0930216 Mfr # TOTE482-ADJ
COMPARE
40 in x 40 in to 48 in x 48 in 160 240
₹648,000.00INR / Each