BriskHeat TOTE-TOP Insulated top cover for adjustable wrap-around tote tank / IBC blanket heaters
Representative image only.
BriskHeat TOTE-TOP Insulated top cover for adjustable wrap-around tote tank / IBC blanket heaters
BriskHeat | Mfr: TOTE-TOP| Item: 0310683
Avoid freezing and increase flow rates of tote tank/IBC contents
₹87,400.00INR / Each
₹87,400.00INR / Each
Product Description
Includes
6-ft power cord with standard 3-prong plug.
Key Features
  • Built-in manual reset high-limit thermostat offers added versatility
  • 1 year warranty
More About this Item
Provide fast uniform heat to your tote tank/IBC contents with this wrap-around blanket heater. The full coverage plug and play system is constructed of a silicone impregnated cloth facing and liner with a 1/4"-thick fiberglass insulation, and is intended for indoor use. Setup and operation is simple — wrap the blanket around your tote tank, secure it with the adjustable nylon straps, and set your desired temperature. Blanket heater will fit any tote tank width from 40"x 40", to 48" x 48".
Not CSA approved
1 Year Warranty
Specifications
  • Drum MaterialTote Tank / IBC
  • Plug TypeNo plug
  • Height (in)36
  • DescriptionInsulated top cover for adjustable wrap-around tote tank / IBC blanket heaters