Branson Stainless Steel Support Racks for Ultrasonic Cleaners
Safely suspends parts off bottom of tank for optimal cleaning
  • Ideal for cleaning small parts Permits sound waves to penetrate directly to the parts
These stainless-steel support racks allow total volume of the ultrasonic cleaner tank to be used while keeping parts, fixtures, or containers about ½" above the tank bottom. By preventing parts from touching the bottom of the tank, the racks eliminate possible damage to the unit and help inhibit ultrasonic activity.
They can also be used as countertop drain racks, holding parts above the countertop surface.
25,600.00 - ₹59,200.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (cm) Length (cm) Width (cm)
Availability Pricing
Branson CPN-916-039 Stainless Steel Support Rack for .5 Gal Ultrasonic Cleaner 0889820 Mfr # CPN-916-039
COMPARE
3.175 14 11.684
₹25,600.00INR / Each
Branson CPN-916-040 Stainless Steel Support Rack for .75 Gal Ultrasonic Cleaner 0889821 Mfr # CPN-916-040
COMPARE
2.032 22.86 12.7
₹27,500.00INR / Each
Branson CPN-916-041 Stainless Steel Support Rack for 1.5 Gal Ultrasonic Cleaner 0889822 Mfr # CPN-916-041
COMPARE
2.032 27.94 13.97
₹35,500.00INR / Each
Branson CPN-916-042 Stainless Steel Support Rack for 2.5 Gal Ultrasonic Cleaner 0889823 Mfr # CPN-916-042
COMPARE
2.032 27.94 21.59
₹39,500.00INR / Each
Branson CPN-916-043 Stainless Steel Support Rack for 5.5 Gal Ultrasonic Cleaner 0889824 Mfr # CPN-916-043
COMPARE
1.905 48.26 27.94
₹59,200.00INR / Each